Learn More
CD164 Antibody (3D7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008763-M11
This item is not returnable.
View return policy
Description
CD164 Monoclonal antibody specifically detects CD164 in Human samples. It is validated for Western Blot, ELISA
Specifications
| CD164 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24v, Multi-glycosylated core protein 24, sialomucin | |
| CD164 (AAH11522.1, 1 a.a. ~ 197 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSRLSRSLLWAATCLGVLCVLSADKNTTQHPNVTTLAPISNVTSAPVTSLPLVTTPAPETCEGRNSCVSCFNVSVVNTTCFWIECKDESYCSHNSTVSDCQVGNTTDFCSVSTATPVPTANSTAKPTVQPSPSTTSKTVTTSGTTNNTVTPTSQPVRKSTFDAASFIGGIVLVLGVQAVIFFLYKFCKSKERNYHTL | |
| 0.1 mg | |
| Cancer | |
| 8763 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 3D7 | |
| Western Blot, ELISA | |
| AAH11522 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |