missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD164 Antibody (502021) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5790JF525
This item is not returnable.
View return policy
Description
CD164 Monoclonal antibody specifically detects CD164 in Human samples. It is validated for Flow Cytometry
Specifications
| CD164 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24v, Multi-glycosylated core protein 24, sialomucin | |
| NS0 mouse myeloma cell line transfected with human CD14, Asp24-Leu197, Accession # Q04900 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 502021 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 8763 | |
| Store at 4°C in the dark. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction