missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD164 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122133
This item is not returnable.
View return policy
Description
CD164 Polyclonal Antibody for Western Blot
This gene encodes a transmembrane sialomucin and cell adhesion molecule that regulates the proliferation, adhesion and migration of hematopoietic progenitor cells. The encoded protein also interacts with the C-X-C chemokine receptor type 4 and may regulate muscle development. Elevated expression of this gene has been observed in human patients with Sezary syndrome, a type of blood cancer, and a mutation in this gene may be associated with impaired hearing.
Specifications
| CD164 | |
| Polyclonal | |
| Unconjugated | |
| CD164 | |
| A115; A24; AA589639; AW540279; Cd164; CD164 antigen; CD164 antigen, sialomucin; CD164 molecule; CD164 molecule, sialomucin; CD164 variant C; c-Myc binding protein; deafness, autosomal dominant 66; DFNA66; endolyn; MGC-24; MGC-24v; MSSP; MUC-24; Multi-glycosylated core protein 24; sialomucin core protein 24 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 83689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9QX82 | |
| CD164 | |
| Recombinant protein Cluster Of Differentiation 164. The antigen corresponds to amino acid range 24-160 of the target protein. | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction