Learn More
Invitrogen™ CD165 Monoclonal Antibody (SN2), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528617
Description
This antibody recognizes an extracellular epitope of CD165, an approximately 37-42 kDa transmembrane glycoprotein expressed mainly on leukemic T cells, double positive and double negative thymocytes (CD4-CD8-, CD4+CD8+), and platelets. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
CD165 (gp37) is a cell surface molecule present on a subset of peripheral lymphocytes and monocytes and is important for adhesion of thymocytes to thymic epithelial cells.
Specifications
| CD165 | |
| Monoclonal | |
| 0.08 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| SN2 | |
| PE | |
| gp137 | |
| CD165, from human Molt-4 cell line. | |
| 100 Tests | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.