missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD166 (ALCAM) Monoclonal Antibody (11-6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA542565
This item is not returnable.
View return policy
Description
Positive Control: PC-3M cell lysate, SHSY5Y cell lysate, A549 cell lysate, rat bladder tissue, human breast cancer tissue, mouse liver tissue, mouse stomach tissue, SHSY5Y. Subcellular Location: Cell membrane, Cell projection, Membrane, Secreted.
This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
| CD166 (ALCAM) | |
| Monoclonal | |
| 2 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O35112, Q13740, Q61490 | |
| ALCAM | |
| Recombinant protein within human CD166 aa 1-300. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 11-6 | |
| Unconjugated | |
| ALCAM | |
| activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
| Mouse | |
| Protein A | |
| RUO | |
| 11658, 214, 79559 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction