missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD166 (ALCAM) Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 159.00 - € 448.00
Specifications
| Antigen | CD166 (ALCAM) |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17730344
![]() |
Invitrogen™
PA5118930 |
200 μL |
€ 448.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17793524
|
Invitrogen™
PA5122131 |
20 μL |
€ 159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD166 (ALCAM) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.Specifications
| CD166 (ALCAM) | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ALCAM | |
| activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
| ALCAM | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O35112, Q13740, Q61490 | |
| 11658, 214, 79559 | |
| Recombinant protein Activated Leukocyte Cell Adhesion Molecule. The antigen corresponds to amino acid range 28-527 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title