missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD169 Monoclonal Antibody (3D6.112)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA516508
This item is not returnable.
View return policy
Description
We recommend using fixation with either 2% paraformaldehyde or ethanol for optimal results. Rat anti Mouse CD169 antibody, clone 3D6.112 recognizes mouse CD169 also known as sialoadhesin, Sheep erythrocyte receptor or Siglec-1.
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
Specifications
| CD169 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q62230 | |
| SIGLEC1 | |
| Purified Sialoadhesin. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| 3D6.112 | |
| Unconjugated | |
| SIGLEC1 | |
| Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN | |
| Rat | |
| Protein G | |
| RUO | |
| 20612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction