missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD169 Recombinant Rabbit Monoclonal Antibody (PSH05-21)

CD169 Recombinant Rabbit Monoclonal Antibody (PSH05-21)
Brand: Invitrogen MA556394
This item is not returnable.
View return policy
Description
CD169 Recombinant Monoclonal Antibody for WB, Flow
Specifications
| CD169 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q62230, Q9BZZ2 | |
| SIGLEC1 | |
| Recombinant protein within human CD169 aa 1-1,000/1,709. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| PSH05-21 | |
| Unconjugated | |
| SIGLEC1 | |
| Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20612, 6614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction