Learn More
Invitrogen™ CD169 (Siglec-1) Monoclonal Antibody (MOMA-1), FITC
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA528189
Product Code. 16372474
  • € 470.00 / 100µg
Description
Rat anti Mouse CD169, clone MOMA-1 recognizes murine CD169, also known as sialoadhesin or Siglec-1. CD169 is a lectin-like receptor expressed by certain populations of macrophages including marginal zone metallophils of the spleen, subcapsular macrophages of lymph nodes and stromal macrophages in bone marrow (Morris et al. 1991).
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD169 (Siglec-1)
Monoclonal
0.1 mg/mL
PBS with 1% BSA and 0.09% sodium azide
Q62230
SIGLEC1
Stromal (reticular) elements from mouse lymph nodes.
100 μg
Primary
Mouse
Antibody
IgG2a
Immunohistochemistry (Frozen), Immunocytochemistry
MOMA-1
FITC
SIGLEC1
Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN
Rat
Protein G
RUO
20612
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.