Learn More
Invitrogen™ CD171 Monoclonal Antibody (L1.1)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ 375100
Product Code. 10342903
  • € 508.00 / 100µg
Description
CD171 Monoclonal Antibody for ICC/IF, IP, ELISA
L1CAM/CD171 is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause three X-linked neurological syndromes known by the acronym CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of a neuron-specific exon is thought to be functionally relevant.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD171
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide; pH 7.4
P32004
L1CAM
3T3 fibroblasts transfected with the human L1 protein.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Immunoprecipitation, Immunocytochemistry
L1.1
Unconjugated
L1CAM
antigen identified by monoclonal antibody R1; CAML1; CD 171; CD171; CD171 molecule; HSAS; HSAS1; Hyd; L1; L1 cell adhesion molecule; L1CAM; MASA; MIC5; N-CAM L1; NCAML1; N-CAML1; N-CAM-L1; NCAM-L1; Nerve-growth factor-inducible large external glycoprotein; neural cell adhesion molecule L1; NILE; S10; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM; SPG1
Mouse
Protein A
RUO
3897
-20°C