Learn More
Invitrogen™ CD171 Monoclonal Antibody (UJ181.4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA146045
Product Code. 11594253
  • € 520.00 / 100µg
Description
CD171 Monoclonal Antibody for Western Blot, IHC, IP
L1CAM/CD171 is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause three X-linked neurological syndromes known by the acronym CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of a neuron-specific exon is thought to be functionally relevant.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD171
Monoclonal
1.0 mg/mL
PBS with 0.02% sodium azide
P32004
L1CAM
16 week human fetal brain.
100 μg
Primary
Human
Antibody
IgG1
Immunohistochemistry, Immunoprecipitation, Western Blot
UJ181.4
Unconjugated
L1CAM
antigen identified by monoclonal antibody R1; CAML1; CD 171; CD171; CD171 molecule; HSAS; HSAS1; Hyd; L1; L1 cell adhesion molecule; L1CAM; MASA; MIC5; N-CAM L1; NCAML1; N-CAML1; N-CAM-L1; NCAM-L1; Nerve-growth factor-inducible large external glycoprotein; neural cell adhesion molecule L1; NILE; S10; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM; SPG1
Mouse
Protein A
RUO
3897
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.