missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD172 Mouse anti-Rat, BV786, Clone: OX-41, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744732
This item is not returnable.
View return policy
Description
CD172 Monoclonal antibody specifically detects CD172 in Rat samples. It is validated for Flow Cytometry.
Specifications
| CD172 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Rat | |
| Purified |
| Flow Cytometry | |
| OX-41 | |
| Brilliant Violet 786 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur