missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD173 Antibody (BRIC231), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62120AF350
This item is not returnable.
View return policy
Description
CD173 Monoclonal specifically detects CD173 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD173 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| NB100-62120 recognises the type 2 H blood group antigen, recently designated CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| BRIC231 | |
| Flow Cytometry | |
| H2, Type 2 H blood group antigen | |
| HEL cell line | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction