missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD177 Antibody (021), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90285H
This item is not returnable.
View return policy
Description
CD177 Monoclonal specifically detects CD177 in Human samples. It is validated for ELISA.
Specifications
| CD177 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 57126 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 21 | |
| ELISA | |
| CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1NB1 glycoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD177 (AAH29167.1; Met1-Gly407). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction