Learn More
CD178 (Fas Ligand) Monoclonal Antibody (NOK-1), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-9919-38
Description
The NOK-1 MAb reacts with human Fas (CD95) Ligand, a 40 kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by neutrophils, monocytes, and activated T cells and NK cells. The interaction of FasL with its receptor (CD95, Fas) induces Fas-mediated killing of lymphocytes. Human FasL is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26 kDa soluble form. Therefore for optimal detection of surface FasL on activated peripheral blood cells, incubation of cells with an MMP inhibitor is recommended.
CD178 (Fas Ligand) Monoclonal antibody specifically detects CD178 (Fas Ligand) in Human samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
Specifications
| CD178 (Fas Ligand) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P48023 | |
| Mouse | |
| 5 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Functional Assay, Neutralization | |
| NOK-1 | |
| Functional Grade | |
| Faslg | |
| ADAM10-processed FasL form; ALPS1B; APL; apoptosis (APO-1) antigen ligand 1; Apoptosis (APO-1) antigen ligand 1 (Fas antigen ligand); apoptosis antigen ligand; APT1LG1; APTL; CD178; CD95 ligand; CD95L; CD95-L; CD95L protein; fas antigen ligand; Fas ligand; Fas ligand (TNF superfamily, member 6); Fasl; Fas-L; FasL ICD; FasL intracellular domain; Faslg; Fas-LG; generalized lymphoproliferative disease; gld; mutant tumor necrosis factor family member 6; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; SPA; SPPL2A-processed FasL form; TNFL; Tnfsf6; TNLG1A; Tumor necrosis factor (ligand) superfamily member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor (ligand) superfamily, member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor ligand; tumor necrosis factor ligand 1A; tumor necrosis factor ligand superfamily member 6; Tumor necrosis factor ligand superfamily member 6, membrane form; Tumor necrosis factor ligand superfamily member 6, soluble form | |
| Affinity chromatography | |
| RUO | |
| 356 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.