missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD180 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534695
This item is not returnable.
View return policy
Description
Recommended positive controls: HL-60. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD180 (RP105, Bgp95, LY64) is a type I membrane glycoprotein of Toll-like receptor (TLR) family. CD180 cytoplasmic tail is short and unlike the TLRs, it lacks the TIR domain. CD180 expression is dependent on the coexpression of its helper molecule, MD-1, and mirrors that of TLR4 on antigen-presenting cells. CD180 regulates recognition of LPS and signaling in B cells, via interacting directly with the TLR4 signaling complex, inhibiting its ability to bind microbial ligands. Ligation of CD180 by monoclonal antibodies leads to B cell activation, upregulation of CD80/CD86, and increase in cell size. CD180 consists of extracellular leucine-rich repeats (LRR) and a short cytoplasmic tail. The extracellular LRR is part of the cell surface receptor complex, RP105/MD-1. Diseases associated with CD180 dysfunction include Kawasaki Disease in children and may play a role in rheumatic disease pathology.
Specifications
| CD180 | |
| Polyclonal | |
| Unconjugated | |
| Cd180 | |
| CD180; CD180 antigen; CD180 molecule; F630107B15; LY64; Ly78; Ly-78; lymphocyte antigen 64; Lymphocyte antigen 78; lymphocyte antigen-64, radioprotective, 105kDa; MGC126233; MGC126234; radioprotective 105 kDa protein; RP105 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4064 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99467 | |
| Cd180 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human RP105. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction