Learn More
Invitrogen™ CD180 (RP105) Monoclonal Antibody (RP/14), PE, eBioscience™

Rat Monoclonal Antibody
Brand: Invitrogen™ 12-1801-81
Description
Description: The RP/14 monoclonal antibody reacts with mouse CD180 (RP105). This 105 kDa type I transmembrane molecule is a member of the TLR family of proteins characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. RP105 physically associates with another molecule called MD-1 and is expressed on mature B cells, monocytes/macrophages and dendritic cells. Ligation of RP105 with RP/14 protects B cells from irradiation- or dexamethasone-induced apoptosis, and drives them to proliferate. B cells activated by RP105-ligation arrest their growth and undergo apoptosis when the antigen receptor is engaged. The RP105/MD-1 complex in concert with TLR4 mediates B cell recognition and signaling of LPS. Thus, RP105 is a signal transduction molecule and plays a role in regulation of B cell growth and death. Applications Reported: The RP/14 antibody has been reported for use in flow cytometric analysis. Applications Tested: The RP/14 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 5...
CD180 (RP105, Bgp95, LY64) is a type I membrane glycoprotein of Toll-like receptor (TLR) family. CD180 cytoplasmic tail is short and unlike the TLRs, it lacks the TIR domain. CD180 expression is dependent on the coexpression of its helper molecule, MD-1, and mirrors that of TLR4 on antigen-presenting cells. CD180 regulates recognition of LPS and signaling in B cells, via interacting directly with the TLR4 signaling complex, inhibiting its ability to bind microbial ligands. Ligation of CD180 by monoclonal antibodies leads to B cell activation, upregulation of CD80/CD86, and increase in cell size. CD180 consists of extracellular leucine-rich repeats (LRR) and a short cytoplasmic tail. The extracellular LRR is part of the cell surface receptor complex, RP105/MD-1. Diseases associated with CD180 dysfunction include Kawasaki Disease in children and may play a role in rheumatic disease pathology.
Specifications
| CD180 (RP105) | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q62192 | |
| Cd180 | |
| Affinity chromatography | |
| RUO | |
| 17079 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| RP/14 | |
| PE | |
| Cd180 | |
| CD180; CD180 antigen; CD180 molecule; F630107B15; LY64; Ly78; Ly-78; lymphocyte antigen 64; Lymphocyte antigen 78; lymphocyte antigen-64, radioprotective, 105kDa; MGC126233; MGC126234; radioprotective 105 kDa protein; RP105 | |
| Rat | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.