missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD183 (CXCR3) Monoclonal Antibody (CEW33D), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 178.00 - € 392.00
Specifications
| Antigen | CD183 (CXCR3) |
|---|---|
| Clone | CEW33D |
| Concentration | 1 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089832
|
Invitrogen™
H087T02B08-A |
25 Tests |
€ 178.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087859
|
Invitrogen™
H087T03B08-A |
100 Tests |
€ 392.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD183 (CXCR3) Monoclonal antibody specifically detects CD183 (CXCR3) in Human samples. It is validated for Flow CytometrySpecifications
| CD183 (CXCR3) | |
| 1 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| an; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD182; CD183; chemokine (C-X-C motif) receptor 3; chemokine receptor 3; CKR-L2; Cmkar3; CXC; C-X-C chemokine receptor type 3; CXC motif chemokine; C-X-C motif chemokine receptor 3; Cxcr3; CXC-R3; CXCR-3; G protein-coupled receptor 9; GPR9; Interferon-inducible protein 10 receptor; IP10; IP10 receptor; IP-10 receptor; IP10-R; Mig receptor; MigR; Mig-R | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| CEW33D | |
| Flow Cytometry | |
| NovaFluor Blue 660-120S | |
| Mouse | |
| Human | |
| P49682 | |
| 2833 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CXCR3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title