missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD185 (CXCR5) Monoclonal Antibody (MU5UBEE), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 178.00 - € 392.00
Specifications
| Antigen | CD185 (CXCR5) |
|---|---|
| Clone | MU5UBEE |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30056088
|
Invitrogen™
H037T03Y03-A |
100 Tests |
€ 392.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055813
|
Invitrogen™
H037T02Y03-A |
25 Tests |
€ 178.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
CD185 (CXCR5) Monoclonal antibody specifically detects CD185 (CXCR5) in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow CytometrySpezifikation
| CD185 (CXCR5) | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human, Monkey, Rhesus Macaque | |
| P32302 | |
| 643, 701792 | |
| IgG2b κ | |
| Affinity chromatography | |
| CXCR5 |
| MU5UBEE | |
| Flow Cytometry | |
| NovaFluor Yellow 610 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Blr1; Blr-1; Burkitt lymphoma receptor 1; burkitt lymphoma receptor 1 homolog; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD185; CD185 antigen; chemochine (C-X-C motif) receptor 5; chemokine (C-X-C motif) receptor 5; CXC; C-X-C chemokine receptor type 5; CXC motif chemokine; C-X-C motif chemokine receptor 5; CXCR5; CXC-R5; CXCR-5; EGK_06966; Fusin; Gpcr6; GPR9; HUMSTR; LESTR; MDR15; MDR-15; Monocyte-derived receptor 15; neurolymphatic receptor; NLR | |
| N-terminal extracellular domain. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts