missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (1D3), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-24965APC
This item is not returnable.
View return policy
Description
CD19 Monoclonal specifically detects CD19 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Cell Surface), Product Image.
Specifications
| CD19 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 0.25 ug/10^6 cells, Flow (Cell Surface) | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Rat | |
| Protein G purified | |
| B Cell Development and Differentiation Markers, Myeloid derived Suppressor Cell | |
| 930 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 1D3 | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CD19 | |
| Mouse CD19 Transfected Cell Line | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction