missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (Bu12), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64360APC
This item is not returnable.
View return policy
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD19 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CD19 | |
| Human EB-4 Burkitt lymphoma cell line. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 930 | |
| Human | |
| Purified |
| Flow Cytometry | |
| Bu12 | |
| Flow Cytometry | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64360 recognizes human CD19, a 95kD cell surface glycoprotein, which is expressed on cells of the B cell lineage and follicular dendritic cells but absent on plasma cells. CD19 is an important signal transduction molecule which is involved in the regulation of B lymphocyte development, activation and differentiation. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction