Learn More
CD19 Antibody (CL15459) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43670-100ul
 View more versions of this product
Product Code. 30279535
  • € 561.00 / 100µL
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD19
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, Immunohistochemistry 1:2500-1:5000, Immunohistochemistry-Paraffin 1:2500-1:5000
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
This antibody was generated using a synthetic peptide of P15391, with the exact immunogen sequence remaining proprietary.
100 μL
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
930
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL15459
Unconjugated
PBS, pH 7.2, 40% glycerol