Learn More
CD19 Antibody (PDR134), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50133-0.025ml
 View more versions of this product
Product Code. 18629236
  • € 188.00 / 0.03mL
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD19
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Pokeweek-stimulated Daudi and Raji cells
0.025 mL
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
930
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgM
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
PDR134
Unconjugated
PBS
Mouse
Thiophilic Chromatography