missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse anti-Human, Monkey, Primate, DyLight 350, Clone: CVID3/429, Novus Biologicals™
Mouse Monoclonal Antibody
€ 615.00
Spezifikation
| Antigen | CD19 |
|---|---|
| Clone | CVID3/429 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | DyLight 350 |
Beschreibung
CD19 Monoclonal specifically detects CD19 in Human, Monkey, Chimpanzee samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Spezifikation
| CD19 | |
| Flow Cytometry, Immunofluorescence | |
| DyLight 350 | |
| Mouse | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 930 | |
| IgG1 κ | |
| Protein A or G purified |
| CVID3/429 | |
| Monoclonal | |
| Purified | |
| RUO | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Recombinant human CD19 protein | |
| Primary | |
| Store at 4°C in the dark. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts