missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse, FITC, Clone: HIB19, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB30-F-25Tests
This item is not returnable.
View return policy
Specifications
| CD19 | |
| Monoclonal | |
| 5μL/Test | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| CD19 | |
| Protein A/G Purified | |
| RUO | |
| 930 | |
| IgG1 κ |
| Flow Cytometry | |
| HIB19 | |
| FITC | |
| NP_001171569 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction