missing translation for 'onlineSavingsMsg'
Learn More

CD19 Mouse, PE, Clone: HIB19, enQuireBio™

Mouse Monoclonal Antibody

€ 85.85 - € 162.00

Specifications

Antigen CD19
Clone HIB19
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15941818
View Documents Promotion Details
EnquireBio
QAB30-PE-25TESTS
25 Tests
€ 85.85
25 tests
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15931818
View Documents Promotion Details
EnquireBio
QAB30-PE-100TESTS
100 Tests
€ 162.00
100 tests
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD19
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
930
IgG1 κ
Protein A/G Purified
HIB19
Flow Cytometry
PE
RUO
NP_001171569
CD19
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications