missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD193 Mouse anti-Human, BV421, Clone: 5E8-G9-B4, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566279
This item is not returnable.
View return policy
Description
CD193 Monoclonal antibody specifically detects CD193 in Human samples. It is validated for Flow cytometry.
Specifications
| CD193 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry | |
| 5E8 (also known as 5E8-G9-B4) | |
| Vol. Per test: 5μL | |
| CCR3: Chemokine (C-C motif) receptor 3; C-C CKR-3; CKR3; CMKBR3 | |
| Human CCR3 Transfected Cell Line | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction