missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD194 Mouse anti-Human, BB700, Clone: 1G1, BD Horizon™
Mouse Monoclonal Antibody
€ 178.71 - € 398.82
Specifications
| Antigen | CD194 |
|---|---|
| Clone | 1G1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15805089
|
BD Biosciences
566476 |
25 Tests |
€ 183.00 € 178.71 / Each Save € 4.29 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15895079
|
BD Biosciences
566475 |
100 Tests |
€ 401.00 € 398.82 / Each Save € 2.18 0.54% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD194 Monoclonal antibody specifically detects CD194 in Human samples. It is validated for Flow Cytometry.Specifications
| CD194 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human | |
| Primary |
| 1G1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title