missing translation for 'onlineSavingsMsg'
Learn More

CD195 Mouse anti-Human, APC, Clone: 2D7/CCR5, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561748

 View more versions of this product

Product Code. 15831299

  • ONLINE EXCLUSIVE PRICE
    € 156.00
    € 154.08 / Each
    Save € 1.92
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 2D7/CCR5 monoclonal antibody specifically binds to the human chemokine receptor, CCR5. CCR5 is also known as CD195. CCR5 is a seven transmembrane-spanning G protein-associated molecule. CCR5 belongs to the beta chemokine receptor family. It is expressed on a subset of T lymphocytes (CD3+CD45RO+CD95+). CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: RANTES and macrophage inflammatory protein-1 (MIP-1), and monocyte chemoattractant protein 2 (MCP-2). Additionally, CCR5 has been found to be a co-receptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele, demonstrate resistance to HIV infection. This antibody has been shown to block ligand and gp120 binding. It is also able to neutralize HIV infection.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD195
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CCR5 Transfected Cell Line
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
2D7/CCR5
CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13
Mouse
Affinity Purified
RUO
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.