missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Antibody (NA1/34-HLK), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64852PECY7
This item is not returnable.
View return policy
Description
CD1a Monoclonal specifically detects CD1a in Human, Canine samples. It is validated for Flow Cytometry.
Specifications
| CD1a | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CD1A | |
| Human thymocytes | |
| 0.1 mL | |
| Primary | |
| NB100-64852 recognizes the human CD1a cell surface glycoprotein, a 49kD molecule expressed in association with beta 2 microglobulin. CD1a is expressed strongly by cortical thymocytes, and also by Langerhans cells and interdigitating cells. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| NA1/34-HLK | |
| Flow Cytometry | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Mouse | |
| Ion exchange chromatography | |
| Dendritic Cell Markers, Immunology | |
| 909.0 | |
| Human, Canine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction