Learn More
CD1a Monoclonal Antibody (HI149), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 323.00
Specifications
Antigen | CD1a |
---|---|
Clone | HI149 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15548326
|
Affymetrix eBioscience
25-0019-42 |
100 Tests |
€ 323.00
100 tests |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The HI149 monoclonal antibody reacts with human CD1a, a 49 kDa protein expressed by cortical thymocytes and dendritic cells including Langerhans cells. The CD1 family of proteins share some structural and functional characteristics with the MHC class I molecules; however, members of the CD1 family are not polymorphic. Similar to MHC class I, CD1a associates with the beta2-microglobulin and is thought to play a role in antigen presentation. Applications Reported: This HI149 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HI149 antibody has been pre-titrated and tested by flow cytometric analysis of MOLT-4 cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Flow Cytometry.Specifications
CD1a | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P06126 | |
CD1A | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD1A |
HI149 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
909 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.