Learn More
CD1a Mouse, anti-Human, Cardinal Red, Clone: HI149, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CR-65172-100TESTS
Description
The CD1a antibody from Proteintech is a mouse monoclonal antibody generated with human thymocytes of human CD1a. This antibody recognizes human antigen. The CD1a antibody has been validated for the following applications: FC analysis. This antibody is cardinal red™.
Specifications
| CD1a | |
| HI149 | |
| Cardinal Red | |
| Mouse | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 5 μL/Test | |
| CD1, CD1A, CD1a molecule, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6 | |
| 100 Tests | |
| 909 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.