missing translation for 'onlineSavingsMsg'
Learn More

CD1a Mouse anti-Human, Unconjugated, Clone: HI149, Proteintech

Product Code. 17264636 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17264636 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17264636 Supplier Proteintech Supplier No. 651721IG100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD1a antibody from Proteintech is a mouse monoclonal antibody to a human thymocytes of human CD1a. This antibody recognizes human antigen. The CD1a antibody has been validated for the following applications: FC analysis.

Specifications

Antigen CD1a
Applications Flow Cytometry
Classification Monoclonal
Clone HI149
Concentration 1 mg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 909
Target Species Human
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.