missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1b Antibody (RIV12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34714DL594
This item is not returnable.
View return policy
Description
CD1b Monoclonal antibody specifically detects CD1b in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications
| CD1b | |
| Monoclonal | |
| DyLight 594 | |
| CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b | |
| Human peripheral lymphocytes (Uniprot: P29016) | |
| 0.1 mL | |
| Immunology | |
| 910 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF | |
| RIV12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu