missing translation for 'onlineSavingsMsg'
Learn More

CD1c/BDCA-1 Antibody (3G1B3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 232.00 - € 612.00

Specifications

Antigen CD1c/BDCA-1
Clone 3G1B3
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18043076
View Documents
Novus Biologicals
NBP2-61725
0.1 mL
€ 612.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18617207
View Documents
Novus Biologicals
NBP2-61725-0.025ml
0.025 mL
€ 232.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD1c/BDCA-1 Monoclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD1c/BDCA-1
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c
CD1C
IgG2b
Protein G purified
3G1B3
Western Blot 1:100-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Monoclonal
Purified
Cancer, Immunology
911
Purified recombinant fragment of human CD1C (AA: extra 18-302) expressed in E. Coli.
Primary
37.7 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.