missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1c/BDCA-1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98701UV
This item is not returnable.
View return policy
Description
CD1c/BDCA-1 Polyclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western Blot
Specifications
| CD1c/BDCA-1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human CD1c/BDCA-1. | |
| 0.1 mL | |
| Cancer, Immunology | |
| 911 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction