missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1c/BDCA-1 Antibody (M241), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2645AF532
This item is not returnable.
View return policy
Description
CD1c/BDCA-1 Monoclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| CD1c/BDCA-1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD1C | |
| MOLT 4 cell line | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| M241 | |
| Immunohistochemistry | |
| BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
| Mouse | |
| Protein A purified | |
| Cancer, Immunology | |
| 911 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction