Learn More
CD1c/BDCA-1 Antibody (M241), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-2645PE
Product Code. 18434799
  • ONLINE EXCLUSIVE PRICE
    € 597.00
    € 563.85 / 0.10mL
    Save € 33.15
    5% Off
Description
CD1c/BDCA-1 Monoclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD1c/BDCA-1
Immunoprecipitation
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c
911