missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1C Monoclonal Antibody (OTI2F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505399S
This item is not returnable.
View return policy
Description
CD1C Monoclonal antibody specifically detects CD1C in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CD1C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P29017 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 911 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI2F1 | |
| Unconjugated | |
| CD1C | |
| BDCA1; canCD1c; CD1; CD1A; CD1C; CD1C antigen, c polypeptide; CD1c molecule; cortical thymocyte antigen CD1C; differentiation antigen CD1-alpha-3; R7; RP11-101J8.3; T-cell surface glycoprotein CD1c | |
| Full length human recombinant protein of human CD1C produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction