missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD1d Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589255

Product Code. 16577211

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EVPQRLFPLR CLQISSFANS SWTRTDGLAW LGELQTHSWS NDSDTVRSLK PWSQGTFSDQ QWETLQHIFR VYRSSFTRDV KEFAKMLRLS YPLELQVSAG CEVHPGNASN NFFHVAFQGK DILSFQGTSW EPTQEAPLWV NLAIQVLNQD KWTRETVQWL LNGTCPQFVS G; Positive Samples: Mouse liver; Cellular Location: Cell membrane, Endosome membrane, Lysosome membrane, Single-pass type I membrane protein.

CD1d is a heavy chain associated with Beta 2 microglobulin on cortical thymocytes. Beta 2 microglobulin independent expression of CD1d has also been demonstrated on human intestinal epithelial cells. CD1d is the sole group 2 member of the CD1 family of major histocompatibility (MHC) like glycoproteins. The CD1d gene encodes a divergent member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by the CD1d gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail. Diseases associated with CD1D include Mycobacterium Malmoense and Autoimmune Disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD1d
Polyclonal
Unconjugated
CD1D
AI747460; Antigen-presenting glycoprotein CD1d; antigen-presenting glycoprotein CD1d1; Cd1; CD1.1; CD1A; Cd1d; CD1D antigen; CD1D antigen, d polypeptide; CD1d molecule; CD1d.1; Cd1d1; CD1d1 antigen; CD1d1 molecule; differentiation antigen CD1-alpha-3; HMC class I antigen-like glycoprotein CD1D; Ly-38; R3; R3G1; T-cell surface glycoprotein CD1d; T-cell surface glycoprotein CD1d1; thymocyte antigen CD1D
Rabbit
Affinity Chromatography
RUO
12479, 912
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Western Blot
1.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11609, P15813
CD1D, Cd1d1
Recombinant fusion protein containing a sequence corresponding to amino acids 20-190 of human CD1D (NP_0017571).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.