missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD1w2 Monoclonal Antibody (CC20)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516617
This item is not returnable.
View return policy
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD1w2 gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to the plasma membrane and to recycling vesicles of the early endocytic system. Alternatively spliced transcript variants have been observed, but their biological validity has not been determined.
Specifications
| CD1w2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| 0 | |
| CD1A | |
| The details of the immunogen for this antibody are not currently available. | |
| 250 μg | |
| Primary | |
| Canine, Bovine, Sheep, Feline, Horse, Goat | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| CC20 | |
| Unconjugated | |
| CD1A | |
| CD1A; CD1a molecule; T-cell surface glycoprotein CD1a | |
| Mouse | |
| Protein A | |
| RUO | |
| 781011 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction