missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (OX-34), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65228B
This item is not returnable.
View return policy
Description
CD2 Monoclonal specifically detects CD2 in Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CD2 | |
| Activated rat T helper cells | |
| 0.25 mL | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 914 | |
| Rat | |
| Purified |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| OX-34 | |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65228 recognizes the rat CD2 cell surface antigen, a 50-54kD glycoprotein expressed by thymocytes and mature T cells. * Whiteland, J.L. et al. (1995). Immunohistochemical Detection of T-Cell Subsets and other Leucocytes in Paraffin-embedded Rat and Mouse Tissues with Monoclonal Antibodies. J. Histochem. Cytochem. 43: 313-320. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction