missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (RPA-2.10), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25200G
This item is not returnable.
View return policy
Description
CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD2 | |
| Monoclonal | |
| DyLight 488 | |
| NP_001758 | |
| CD2 | |
| A recombinant protein was used as the immunogen for the CD2 antibody. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 914 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| RPA-2.10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction