missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (TS1/8), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37714JF646
This item is not returnable.
View return policy
Description
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Flow Cytometry, CyTOF, Neutralization
Specifications
| CD2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF, Neutralization | |
| TS1/8 | |
| Flow Cytometry, CyTOF-ready, Block/Neutralize | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Cytotoxic T lymphocytes. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells | |
| 914 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction