missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD2 Monoclonal Antibody (RM2-5), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

€ 176.00 - € 339.00

Specifications

Antigen CD2
Clone RM2-5
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088505
View Documents
Invitrogen™
M058T02Y03-A
25 μg
€ 176.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088005
View Documents
Invitrogen™
M058T03Y03-A
100 μg
€ 339.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD2 Monoclonal antibody specifically detects CD2 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD2
0.1 mg/mL
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
Cd2; CD2 antigen; CD2 antigen (p50), sheep red blood cell receptor; CD2 molecule; CD2R; cluster of differentiation 2; Erythrocyte receptor; FLJ46032; LFA2; LFA-2; LFA-3 receptor; Ly37; Ly-37; lymphocyte antigen 37; OX34; OX-34; OX-34 antigen; OX-45 surface antigen homolog to human T lymphocyte CD2 antigen; Rosette receptor; SRBC; surface immunoglobulin superfamily molecule; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5
IgG2b λ
Affinity chromatography
Antibody
RM2-5
Flow Cytometry
NovaFluor Yellow 610
Rat
Mouse
P08920
12481
Primary
4°C, store in dark, DO NOT FREEZE!
CD2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.