Learn More
CD2 Mouse, anti-Human, PerCP, Clone: RPA-2.10, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 100-0428
This item is not returnable.
View return policy
Description
- The RPA-2.10 antibody reacts with CD2, an ∼50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion
- CD2 is broadly expressed on peripheral T and NK cells, dendritic cells, erythrocytes, most thymic T cells, subsets of thymic B cells, and on the endothelium
- CD2 expression appears early during T cell differentiation
- Aberrant expression has been observed in some lymphomas and myeloid leukemias
- CD2 is critically important for T cell activation and signaling, as well as lymphocyte adhesion
- The primary ligand for CD2 is CD58 (LFA-3) located on antigen-presenting cells, with additional ligands comprising CD15 (SSEA-1), CD48 and CD59
- Notably, the RPA-2.10 antibody blocks the mixed lymphocyte reaction
Specifications
| CD2 | |
| Monoclonal | |
| PerCP | |
| 25 Tests | |
| Baboon, Cynomolgus, Pig, Human |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| RPA-2.10 | |
| Mouse | |
| Primary | |
| T Cells |