Learn More
CD2 Mouse, anti-Human, PerCP, Clone: RPA-2.10, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 100-0428
Encompass_Preferred
 View more versions of this product
Product Code. 17167591
  • € 171.45 / 25 tests
Description
  • The RPA-2.10 antibody reacts with CD2, an ∼50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion
  • CD2 is broadly expressed on peripheral T and NK cells, dendritic cells, erythrocytes, most thymic T cells, subsets of thymic B cells, and on the endothelium
  • CD2 expression appears early during T cell differentiation
  • Aberrant expression has been observed in some lymphomas and myeloid leukemias
  • CD2 is critically important for T cell activation and signaling, as well as lymphocyte adhesion
  • The primary ligand for CD2 is CD58 (LFA-3) located on antigen-presenting cells, with additional ligands comprising CD15 (SSEA-1), CD48 and CD59
  • Notably, the RPA-2.10 antibody blocks the mixed lymphocyte reaction
Specifications
Specifications
CD2
Monoclonal
PerCP
25 Tests
Baboon, Cynomolgus, Pig, Human
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
RPA-2.10
Mouse
Primary
T Cells