Learn More
CD2 Mouse, PE, Clone: RPA-2.10, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio QAB1-PE-100Tests
 View more versions of this product
Product Code. 15911468
  • € 433.65 / 100 tests
Specifications
CD2
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD2
Protein A/G Purified
RUO
914
IgG1 κ
Flow Cytometry
RPA-2.10
PE
NP_001758
Mouse
100 Tests
Primary
Store at 2-4°C.