Learn More
CD20 Antibody (2709D), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB42252M
Product Code. 30060186
  • € 619.00 / 0.10mL
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD20
Monoclonal
DyLight 594
50mM Sodium Borate
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunohistochemistry
2709D
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Human CD20 Peptide. The epitope recognized by rituximab, Binder M, et al. (2006) 108:1975-8. doi: 10.1182/blood-2006-04-014639. Epub 2006 May 16. PMID: 16705086
0.1 mL
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
931
Store at 4C in the dark.