missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (2H7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62206F
This item is not returnable.
View return policy
Description
CD20 Monoclonal specifically detects CD20 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
| CD20 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MS4A1 | |
| Human tonsillar B cells | |
| 0.1 mL | |
| Primary | |
| The antibody MFG-06 reacts with a 40-45 kDa glycoprotein in human milk fat globule membrane preparation. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| 2H7 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Human, Primate | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction