missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (AISB12), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43435MFV500
This item is not returnable.
View return policy
Description
CD20 Monoclonal antibody specifically detects CD20 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| CD20 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| AISB12 | |
| Flow Cytometry | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| The immunogen for this antibody was CD20. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction